tert-Butyl-4-amino-4-(4-bromophenyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-amino-4-(4-bromophenyl)piperidine-1-carboxylate

A105141-1G A105141-250MG
76809-534EA 294.94 USD
76809-534 76809-536
tert-Butyl-4-amino-4-(4-bromophenyl)piperidine-1-carboxylate 97%
tert-Butyl-4-amino-4-(4-bromophenyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₃BrN₂O₂
MW: 355.28 g/mol
MDL Number: MFCD11847939
CAS Number: 917925-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR