methyl 5-methoxypyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

A104500-100MG A104500-1G A104500-250MG A104500-5G
76809-492EA 9 USD
76809-492 76809-494 76809-496 76809-498
methyl 5-methoxypyridine-2-carboxylate 98%
methyl 5-methoxypyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD11975090
CAS Number: 29681-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR