Methyl 2,6-dimethylisonicotinate 95%

Supplier: Ambeed
Warning

A104456-100MG A104456-1G A104456-250MG A104456-5G
76809-484EA 6 USD
76809-484 76809-486 76809-488 76809-490
Methyl 2,6-dimethylisonicotinate 95%
Methyl 2,6-dimethylisonicotinate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MDL Number: MFCD11656351
CAS Number: 142896-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR