2,4'-Dichlorobenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: 2,4'-DBP, (2-Chlorophenyl)(4-chlorophenyl)methanone

A104037-100G A104037-10G A104037-25G A104037-500G
76809-456EA 39.26 USD
76809-456 76809-458 76809-460 76809-462
2,4'-Dichlorobenzophenone 98%
2,4'-Dichlorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₈Cl₂O
MW: 251.11 g/mol
MDL Number: MFCD00038744
CAS Number: 85-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR