Spiro-OMeTAD 98%

Supplier: Ambeed
Warning

Synonyms: 2,2',7,7'-Tetrakis[N,N-di(4-methoxyphenyl)amino]-9,9'-spirobifluorene, N2,N2,N2',N2',N7,N7,N7',N7'-Octakis(4-methoxyphenyl)-9,9'-spirobi[fluorene]-2,2',7,7'-tetraamine

A103973-100MG A103973-1G A103973-250MG
76809-448EA 37.14 USD
76809-448 76809-450 76809-452
Spiro-OMeTAD 98%
Spiro-OMeTAD

Precaución: for Research Use Only

Formula: C₈₁H₆₈N₄O₈
MW: 1225.42 g/mol
MDL Number: MFCD12022511
CAS Number: 207739-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR