2-Chloro-7-methoxyquinoxaline 95%

Supplier: Ambeed
Warning

A103566-100MG A103566-250MG
76809-398EA 57.29 USD
76809-398 76809-400
2-Chloro-7-methoxyquinoxaline 95%
2-Chloro-7-methoxyquinoxaline

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O
MW: 194.6200 g/mol
MDL Number: MFCD11846475
CAS Number: 55686-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR