Methyl-4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(2-(2-amino-4-oxo-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzoate, 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid Methyl Ester, 2-Amino-4,7-dihydro-5-[2-[4-(methoxycarbonyl)phenyl]ethyl]-4-oxo-3H-pyrrolo[2,3-d]pyrimidine

A102666-10G A102666-250MG A102666-5G A102666-1G
76809-348EA 36 USD
76809-348 76809-350 76809-352 77570-154
Methyl-4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate 97%
Methyl-4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₄O₃
MW: 312.33 g/mol
MDL Number: MFCD17976681
CAS Number: 155405-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR