(S)-1-((Benzyloxy)carbonyl)hexahydropyridazine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A102545-250MG A102545-1G A102545-5G
76809-334EA 41.38 USD
76809-334 77670-476 77670-478
(S)-1-((Benzyloxy)carbonyl)hexahydropyridazine-3-carboxylic acid 95%
(S)-1-((Benzyloxy)carbonyl)hexahydropyridazine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₄
MW: 264.2771 g/mol
MDL Number: MFCD09751404
CAS Number: 65632-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR