Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Gly-OEt.HCl, Glycine Ethyl Ester HCl, Ethyl Glycinate Hydrochloride
A102317-1000G
A102317-100G
A102317-25G
A102317-500G
A102317-5G
A102317-10G
76809-296EA
31.83
USD
76809-296
76809-298
76809-300
76809-302
76809-304
77592-080
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) 98%
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl)
Precaución: for Research Use Only
Formula:
C₄H₁₀ClNO₂ MW: 139.58 g/mol |
MDL Number:
MFCD00012871 CAS Number: 623-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...