Potassium 6-Hydroxy-2-naphthalenesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Schaeffer's acid potassium salt, 6-Hydroxy-2-naphthalenesulphonic acid potassium salt, Potassium 2-Naphthol-6-sulphonate, 2-Naphthol-6-sulphonic acid potassium salt

A998709-25G A998709-500G A998709-100G
76808-218EA 14 USD
76808-218 77880-819 77880-818
Potassium 6-Hydroxy-2-naphthalenesulfonate 98%
Potassium 6-Hydroxy-2-naphthalenesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₈O₄S
MW: 262.32 g/mol
MDL Number: MFCD06797149
CAS Number: 833-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR