(1R,2R,4S)-2-((R)-1,2-Dihydroxyethyl)-3,6-diphenylbicyclo[2.2.0]hexane-1,2,4,5-tetraol 98%

Supplier: Ambeed
Warning

A995626-1G A995626-5G
76808-214EA 9 USD
76808-214 76808-216
(1R,2R,4S)-2-((R)-1,2-Dihydroxyethyl)-3,6-diphenylbicyclo[2.2.0]hexane-1,2,4,5-tetraol 98%
(1R,2R,4S)-2-((R)-1,2-Dihydroxyethyl)-3,6-diphenylbicyclo[2.2.0]hexane-1,2,4,5-tetraol

Precaución: for Research Use Only

Formula: C₂₀H₂₂O₆
CAS Number: 2845120-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR