(R)-(-)-Ethyl mandelate 98%

Supplier: Ambeed
Warning

A983882-25G A983882-5G
76808-200EA 21 USD
76808-200 76808-202
(R)-(-)-Ethyl mandelate 98%
(R)-(-)-Ethyl mandelate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00064248
CAS Number: 10606-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR