N,N-Dimethyl-2-phenoxyethanamine 98%
Supplier: Ambeed
|
Warning
|
A983531-100G
A983531-10G
A983531-1G
A983531-25G
A983531-500G
A983531-5G
76808-188EA
485.01
USD
76808-188
76808-190
76808-192
76808-194
76808-196
76808-198
N,N-Dimethyl-2-phenoxyethanamine 98%
N,N-Dimethyl-2-phenoxyethanamine
Precaución: for Research Use Only
Formula:
C₁₀H₁₅NO |
MDL Number:
MFCD00040016 CAS Number: 13468-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...