1-Acetyl-2-naphthol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Acetyl-2-hydroxynaphthalene, 2-Hydroxy-1-acetonaphthone, 1-(2-Hydroxynaphthalen-1-yl)ethanone, 2'-Hydroxy-1'-acetonaphthone, 1-(2-Hydroxy-1-naphthyl)ethan-1-one

A978998-100G A978998-25G A978998-5G
76808-176EA 203.7 USD
76808-176 76808-178 76808-180
1-Acetyl-2-naphthol 98%
1-Acetyl-2-naphthol

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
MDL Number: MFCD00012131
CAS Number: 574-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR