2-(2-Ethoxyethoxy)benzamide 98%

Supplier: Ambeed
Warning

A973390-10MG A973390-1MG
76808-172EA 8 USD
76808-172 77670-416
2-(2-Ethoxyethoxy)benzamide 98%
2-(2-Ethoxyethoxy)benzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MDL Number: MFCD00864367
CAS Number: 15302-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR