Imidazo[1,2-a]pyridin-2-ylmethanol 90%

Supplier: Ambeed
Warning

Synonyms: 2-(Hydroxymethyl)imidazo[1,2-a]pyridine

A970240-100MG A970240-1G A970240-250MG A970240-5G
76808-156EA 20.16 USD
76808-156 76808-158 76808-160 76808-162
Imidazo[1,2-a]pyridin-2-ylmethanol 90%
Imidazo[1,2-a]pyridin-2-ylmethanol

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.17 g/mol
MDL Number: MFCD06797471
CAS Number: 82090-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR