Diethyl itaconate 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A962201-100G A962201-1G A962201-25G A962201-5G
76808-118EA 63 USD
76808-118 76808-120 76808-122 76808-124
Diethyl itaconate 98% stabilized
Diethyl itaconate

Precaución: for Research Use Only

Formula: C₉H₁₄O₄
MDL Number: MFCD00059230
CAS Number: 2409-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR