Ethylenebis(triphenylphosphine)platinum(0) 98%

Supplier: Ambeed
Warning

Synonyms: ethene, platinum, triphenylphosphane

A951258-100MG A951258-1G A951258-250MG
76808-078EA 79 USD
76808-078 76808-080 76808-082
Ethylenebis(triphenylphosphine)platinum(0) 98%
Ethylenebis(triphenylphosphine)platinum(0)

Precaución: for Research Use Only

Formula: C₃₈H₃₄P₂Pt
MW: 747.722 g/mol
MDL Number: MFCD00012028
CAS Number: 12120-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR