Ethoxy(methyl)diphenylsilane 95%

Supplier: Ambeed
Warning

A948684-10G A948684-1G A948684-250MG A948684-5G
76808-070EA 268.41 USD
76808-070 76808-072 76808-074 76808-076
Ethoxy(methyl)diphenylsilane 95%
Ethoxy(methyl)diphenylsilane

Precaución: for Research Use Only

Formula: C₁₅H₁₈OSi
MW: 242.39 g/mol
MDL Number: MFCD00026752
CAS Number: 1825-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR