Tri-p-tolylamine 98%

Supplier: Ambeed
Warning

Synonyms: 4,4',4''-Trimethyltriphenylamine

A947509-100G A947509-1G A947509-25G A947509-5G
76808-062EA 108.22 USD
76808-062 76808-064 76808-066 76808-068
Tri-p-tolylamine 98%
Tri-p-tolylamine

Precaución: for Research Use Only

Formula: C₂₁H₂₁N
MW: 287.41 g/mol
MDL Number: MFCD00674043
CAS Number: 1159-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR