Iron(II) oxalate hydrate Fe> 30.5%

Supplier: Ambeed
Warning

A941330-1000G A941330-100G A941330-500G
76808-056EA 24 USD
76808-056 76808-058 76808-060
Iron(II) oxalate hydrate Fe> 30.5%
Iron(II) oxalate hydrate

Precaución: for Research Use Only

Formula: FeC₂O₄·xH₂O
MDL Number: MFCD00012475
CAS Number: 516-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR