(S)-3-(Aminomethyl)-1-boc-piperidine 98%

Supplier: Ambeed
Warning

A930037-1G A930037-5G A930037-25G
76808-048EA 33 USD
76808-048 77595-506 77880-757
(S)-3-(Aminomethyl)-1-boc-piperidine 98%
(S)-3-(Aminomethyl)-1-Boc-piperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD03839877
CAS Number: 140645-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR