Diethyl 1-benzylpyrrolidine-2,5-dicarboxylate 98%

Supplier: Ambeed
Warning

A928946-1G A928946-250MG A928946-5G
76808-042EA 114 USD
76808-042 76808-044 76808-046
Diethyl 1-benzylpyrrolidine-2,5-dicarboxylate 98%
Diethyl 1-benzylpyrrolidine-2,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₄
MDL Number: MFCD00830964
CAS Number: 17740-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR