H-D-Ser -OHHCL 97%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Acetoxy-2-aminopropanoic acid hydrochloride, H-D-Ser(Ac)-OH·HCl

A928580-100MG A928580-1G A928580-250MG
76808-036EA 28.65 USD
76808-036 76808-038 76808-040
H-D-Ser -OHHCL 97%
H-D-Ser -OHHCL

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO₄
MW: 183.588 g/mol
MDL Number: MFCD00672365
CAS Number: 201212-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR