Ethyl-2-aminobenzo[b]thiophene-3-carboxylate 98%

Supplier: Ambeed
Warning

A925018-100MG A925018-250MG
76808-024EA 98.67 USD
76808-024 76808-026
Ethyl-2-aminobenzo[b]thiophene-3-carboxylate 98%
Ethyl 2-aminobenzo[b]thiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂S
MDL Number: MFCD00102506
CAS Number: 7311-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR