Fluoromethyl phenyl sulfone 98%

Supplier: Ambeed
Warning

Synonyms: Fluoromethylsulphonylbenzene, ((Fluoromethyl)sulphonyl)benzene

A913268-1G A913268-250MG A913268-5G
76807-990EA 60.48 USD
76807-990 76807-992 76807-994
Fluoromethyl phenyl sulfone 98%
Fluoromethyl phenyl sulfone

Precaución: for Research Use Only

Formula: C₇H₇FO₂S
MW: 174.19 g/mol
MDL Number: MFCD00191650
CAS Number: 20808-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR