(S)-Ethyl 2-amino-3-(4-chlorophenyl)propanoate hydrochloride 95+%

Supplier: Ambeed
Warning

A911715-250MG A911715-5G A911715-1G
76807-984EA 17 USD
76807-984 76807-986 77880-737
(S)-Ethyl 2-amino-3-(4-chlorophenyl)propanoate hydrochloride 95+%
(S)-Ethyl 2-amino-3-(4-chlorophenyl)propanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅Cl₂NO₂
MDL Number: MFCD00012531
CAS Number: 51366-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR