Ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate

A910667-100G A910667-10G A910667-1G A910667-25G A910667-5G
76807-970EA 205 USD
76807-970 76807-972 76807-974 76807-976 76807-978
Ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate 97%
Ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂S
MDL Number: MFCD00085051
CAS Number: 4815-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR