N-(2-Chlorobenzyloxycarbonyloxy)succinimide 97%

Supplier: Ambeed
Warning

Synonyms: O-(2-Chlorobenzyloxycarbonyl)-N-hydroxysuccinimide, 2-Chlorobenzyl (2,5-dioxopyrrolidin-1-yl) carbonate, 2-Chlorobenzyl Succinimidyl Carbonate, N-(2-Chlorocarbobenzoxyoxy)succinimide

A905938-100G A905938-25G A905938-5G
76807-960EA 133.68 USD
76807-960 76807-962 77670-334
N-(2-Chlorobenzyloxycarbonyloxy)succinimide 97%
N-(2-Chlorobenzyloxycarbonyloxy)succinimide

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO₅
MW: 283.66 g/mol
MDL Number: MFCD00010743
CAS Number: 65853-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR