N-(2-Cyanoethyl)-N-ethylaniline 98+%

Supplier: Ambeed
Warning

Synonyms: 3-(Ethyl(phenyl)amino)propanenitrile, 3-(N-Ethyl-N-phenylamino)propionitrile

A905916-100G A905916-25G A905916-500G
76807-954EA 18 USD
76807-954 76807-956 76807-958
N-(2-Cyanoethyl)-N-ethylaniline 98+%
N-(2-Cyanoethyl)-N-ethylaniline

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂
MDL Number: MFCD00019858
CAS Number: 148-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR