6-Hydroxy-1-naphthoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Hydroxy-1-naphthalenecarboxylic acid
A901379-100G
A901379-10G
A901379-1G
A901379-250MG
A901379-25G
A901379-5G
76807-940EA
581.38
USD
76807-940
76807-942
76807-944
76807-946
76807-948
76807-950
6-Hydroxy-1-naphthoic acid 98%
6-Hydroxy-1-naphthoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₈O₃ MW: 188.1800 g/mol |
MDL Number:
MFCD00209983 CAS Number: 2437-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...