N,N-Diisopropylcarbamoyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: Diisopropylcarbamoyl Chloride

A899940-25G A899940-5G
76807-936EA 97 USD
76807-936 77569-626
N,N-Diisopropylcarbamoyl chloride 98%
N,N-Diisopropylcarbamoyl chloride

Precaución: For research use only.

Formula: C₇H₁₄ClNO
MW: 163.6452 g/mol
MDL Number: MFCD00015013
CAS Number: 19009-39-3
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR