Dimethylphenylsilane 98%

Supplier: Ambeed
Danger

Synonyms: Dimethyl(phenyl)silane

A899218-25G A899218-5G
76807-924EA 38.2 USD
76807-924 76807-926
Dimethylphenylsilane 98%
Dimethylphenylsilane

Precaución: for Research Use Only

Formula: C₈H₁₂Si
MW: 136.27 g/mol
MDL Number: MFCD00008257
CAS Number: 766-77-8
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR