Didodecylamine 97% (by GC)

Supplier: Ambeed
Warning

A894131-1G A894131-5G
76807-914EA 9 USD
76807-914 76807-916
Didodecylamine 97% (by GC)
Didodecylamine

Precaución: for Research Use Only

Formula: C₂₄H₅₁N
MDL Number: MFCD00041918
CAS Number: 3007-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR