1,4-Bis[(9S)-10,11-dihydro-6′-methoxycinchonan-9-yl]-9,10-anthracenedione 98%

Supplier: Ambeed
Warning

A893134-100MG A893134-1G A893134-250MG A893134-5G
76807-910EA 91.24 USD
76807-910 76807-912 77603-234 77670-304
1,4-Bis[(9S)-10,11-dihydro-6′-methoxycinchonan-9-yl]-9,10-anthracenedione 98%
1,4-Bis[(9S)-10,11-dihydro-6′-methoxycinchonan-9-yl]-9,10-anthracenedione

Precaución: for Research Use Only

Formula: C₅₄H₅₆N₄O₆
MW: 857.04 g/mol
MDL Number: MFCD29075644
CAS Number: 176298-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR