1,4-Bis(2-hydroxyethoxy)benzene 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Hydroquinone Di(2-hydroxyethyl) Ether, 1,4-Di(2-hydroxyethoxy)benzene, Hydroquinone Bis(2-hydroxyethyl) Ether
A884586-100G
A884586-25G
A884586-500G
76807-880EA
18.04
USD
76807-880
76807-882
76807-884
1,4-Bis(2-hydroxyethoxy)benzene 95%
1,4-Bis(2-hydroxyethoxy)benzene
Precaución: for Research Use Only
Formula:
C₁₀H₁₄O₄ MW: 198.22 g/mol |
MDL Number:
MFCD00002861 CAS Number: 104-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...