Isoquinoline-5-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 5-Cyanoisoquinoline

A884017-100MG A884017-250MG A884017-5G
76807-874EA 20.16 USD
76807-874 76807-876 76807-878
Isoquinoline-5-carbonitrile 97%
Isoquinoline-5-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆N₂
MW: 154.1700 g/mol
MDL Number: MFCD03109890
CAS Number: 27655-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR