Methyl imidazo[1,2-a]pyrazine-8-carboxylate 95%

Supplier: Ambeed
Warning

A879921-250MG A879921-5G A879921-1G
76807-842EA 30.77 USD
76807-842 76807-844 77603-200
Methyl imidazo[1,2-a]pyrazine-8-carboxylate 95%
Methyl imidazo[1,2-a]pyrazine-8-carboxylate

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD07779193
CAS Number: 850349-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR