N-(Piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide 95%

Supplier: Ambeed
Warning

Synonyms: Flecainide

A879202-25MG A879202-50MG
76807-838EA 8.49 USD
76807-838 76807-840
N-(Piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide 95%
N-(Piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide

Precaución: for Research Use Only

Formula: C₁₇H₂₀F₆N₂O₃
MW: 414.34 g/mol
MDL Number: MFCD00864713
CAS Number: 54143-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR