Ethyl-3-(2-thienyl)-1H-pyrazole-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-(2-Thienyl)pyrazole-3-carboxylate

A878616-1G A878616-5G
76807-834EA 22.28 USD
76807-834 76807-836
Ethyl-3-(2-thienyl)-1H-pyrazole-5-carboxylate 97%
Ethyl-3-(2-thienyl)-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂S
MW: 222.262 g/mol
MDL Number: MFCD00099767
CAS Number: 121195-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR