Ethyl-2-phenyl-2-cyanobutylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-cyano-2-phenylbutanoate

A878245-100G A878245-10G A878245-1G A878245-25G A878245-5G
76807-824EA 117.76 USD
76807-824 76807-826 76807-828 76807-830 76807-832
Ethyl-2-phenyl-2-cyanobutylate 98%
Ethyl-2-phenyl-2-cyanobutylate

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₂
MW: 217.268 g/mol
MDL Number: MFCD00068977
CAS Number: 718-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR