2-(2-Chlorophenyl)-3-methyl-5-pyrazolone 98%

Supplier: Ambeed
Warning

A876683-1000G A876683-100G A876683-25G A876683-500G
76807-816EA 130 USD
76807-816 76807-818 76807-820 76807-822
2-(2-Chlorophenyl)-3-methyl-5-pyrazolone 98%
2-(2-Chlorophenyl)-3-methyl-5-pyrazolone

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O
MDL Number: MFCD00043809
CAS Number: 14580-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR