4-Amino-8-(trifluoromethyl)quinoline 97%

Supplier: Ambeed
Warning

Synonyms: 8-(Trifluoromethyl)quinolin-4-amine

A875831-100MG A875831-1G
76807-812EA 22.28 USD
76807-812 76807-814
4-Amino-8-(trifluoromethyl)quinoline 97%
4-Amino-8-(trifluoromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₇F₃N₂
MW: 212.18 g/mol
MDL Number: MFCD00269346
CAS Number: 243977-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR