Ethyl-5-hydroxy-2-methyl-1H-indole-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-2-methylindole-3-carboxylic acid ethyl ester, Ethyl 5-hydroxy-2-methyl-1H-indole-3-carboxylate

A875543-1G A875543-5G
76807-810EA 45.62 USD
76807-810 77670-266
Ethyl-5-hydroxy-2-methyl-1H-indole-3-carboxylate 97%
Ethyl-5-hydroxy-2-methyl-1H-indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
MW: 219.24 g/mol
MDL Number: MFCD00044575
CAS Number: 7598-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR