Isoquinoline-1-carbonitrile 95%

Supplier: Ambeed
Warning

A875423-100G A875423-1G A875423-250MG A875423-25G A875423-5G
76807-800EA 1318 USD
76807-800 76807-802 76807-804 76807-806 76807-808
Isoquinoline-1-carbonitrile 95%
Isoquinoline-1-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆N₂
MDL Number: MFCD00134166
CAS Number: 1198-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR