H-D-Lys(boc)-OtBu·HCl 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Lys(Boc)-OtBu.HCl, H-D-Lys(Boc)-OtBu·HCl

A874332-100MG A874332-1G A874332-25G
76807-794EA 28.65 USD
76807-794 76807-796 76807-798
H-D-Lys(boc)-OtBu·HCl 98%
H-D-Lys(boc)-OtBu·HCl

Precaución: for Research Use Only

Formula: C₁₅H₃₁ClN₂O₄
MW: 338.873 g/mol
MDL Number: MFCD00237143
CAS Number: 201007-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR