Dimethyl 2-nitroisophthalate 97%

Supplier: Ambeed
Warning

A873989-100MG A873989-1G A873989-250MG A873989-5G
76807-786EA 9.55 USD
76807-786 76807-788 76807-790 76807-792
Dimethyl 2-nitroisophthalate 97%
Dimethyl 2-nitroisophthalate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₆
MW: 239.18 g/mol
MDL Number: MFCD18651700
CAS Number: 57052-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR