1-Bromo-4-fluoronaphthalene 95%

Supplier: Ambeed
Warning

A873560-100G A873560-10G A873560-1G A873560-25G A873560-5G
76807-776EA 91.24 USD
76807-776 76807-778 76807-780 76807-782 76807-784
1-Bromo-4-fluoronaphthalene 95%
1-Bromo-4-fluoronaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆BrF
MW: 225.06 g/mol
MDL Number: MFCD00051473
CAS Number: 341-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR