4-Chlorobutyrophenone 95%

Supplier: Ambeed
Warning

Synonyms: ω-Chlorobutyrophenone, γ-Chlorobutyrophenone

A870859-100G A870859-25G A870859-5G
76807-766EA 432.85 USD
76807-766 76807-768 76807-770
4-Chlorobutyrophenone 95%
4-Chlorobutyrophenone

Precaución: for Research Use Only

Formula: (C₆H₅)C(O)(CH₂)₃Cl
MW: 182.65 g/mol
MDL Number: MFCD00001005
CAS Number: 939-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR