Ethyl-5-chloronicotinate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-chloro-5-pyridinecarboxylate

A869416-250MG A869416-25G A869416-5G
76807-760EA 22.28 USD
76807-760 76807-762 76807-764
Ethyl-5-chloronicotinate 96%
Ethyl-5-chloronicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
MDL Number: MFCD01692171
CAS Number: 20825-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR